Learn More
Research Products International Corp Bile Salts Number 3, 100 Grams
SDP
Supplier:  Research Products International Corp B11750100.0
Encompass_Preferred
Bile Salts No. 3 is refined bile salt, comprising mainly of sodium cholate and sodium deoxycholate. It is used in media as a agent to isolate enteric organisms and coliforms. It is an excellent inhibitor of gram positive bacteria such as streptococci and staphylocci.
Catalog No. 50-995-793