Learn More
Apexbio Technology LLC Bilirubin 635-65-4 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC N25845.1
Encompass_Preferred
Bilirubin (CAS 635-65-4) is a bile pigment naturally produced from the catabolism of heme It exhibits antioxidant properties primarily via scavenging reactive oxygen species and inhibiting oxidative modification of biomolecules Bilirubin modulates signaling pathways associated with inflammation oxidative stress and cellular apoptosis rendering it relevant in biomedical research focused on hepatic disorders cardiovascular disease and neurodegenerative conditions Originally identified from bezoar bilirubin requires storage under airtight cool and dry conditions to maintain stability
Catalog No. 50-000-63495
 Restricted Item