Learn More
Medchemexpress LLC Biliverdin hydrochloride | 856699-18-8 | MFCD06245593 | 99.3% | 619.11 g/mol | C33H35ClN4O6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1350055MG
Encompass_Preferred
Biliverdin hydrochloride is a water-soluble tetrapyrrolic bile pigment produced during heme catabolism. Supplied as a green to dark-green solid, it is used as a biochemical reagent in studies of heme metabolism, oxidative stress, and inflammation. Analytical identifiers include CAS 856699-18-8, formula C33H35ClN4O6, and molecular weight 619.11 g/mol.
  • High purity (99.3%) suitable for in vitro biochemical applications.
  • Available in small research pack sizes, including 5 mg, for assay development.
  • Soluble in DMSO (20 mg/mL) with warming or sonication; low water solubility.
  • Store sealed, away from moisture; stable at -20°C, with longer stability at -80°C in solution.
  • Green to dark green solid allowing easy visual confirmation of material.
Catalog No. 50-002-22050