Learn More
Selleck Chemical LLC Bilobalide 10mg 33570-04-6
SDP
Supplier:  Selleck Chemical LLC S227610MG
Encompass_Preferred
Bilobalide is a biologically active terpenic trilactone present in Ginkgo biloba. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-775-0