Learn More
Abcam BIM 23052, Somatostatin (sst5) receptor 5 agonist, 1MG
Supplier:  Abcam AB1412131MG
Encompass_Preferred
MW 1122.33 g/mol. Selective somatostatin (sst₅) receptor 5 agonist (Ki values are 7.3 (sst₅), 13.5 (sst₂), 31.3 (sst₁) and 141 nM (sst₄)). Role in gastrointestinal and neuroendocrine systems. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6793