Learn More
TARGETMOL CHEMICALS INC BIMIRALISIB 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T226510MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Bimiralisib (PI3K-IN-2) is an orally bioavailable pan inhibitor of PI3K and inhibitor of the mTOR with potential antineoplastic activity. PI3K-IN-2 inhibits the PI3K kinase isoforms alpha beta gamma and delta and to a lesser extent mTOR kinase which may result in tumor cell apoptosis and growth inhibition in cells overexpressing PI3K/mTOR. Activation of the PI3K/mTOR pathway promotes cell growth survival and resistance to both chemotherapy and radiotherapy. purity: 99%
Catalog No. 50-257-7185