Learn More
Selleck Chemical LLC Bimiralisib (PQR309) 5mg 1225037-39-7
SDP
Supplier:  Selleck Chemical LLC S87385MG
Encompass_Preferred
Bimiralisib (PQR309) is a novel brain-penetrant dual PI3K/mTOR inhibitor with in vitro and in vivo antilymphoma activity. It displays excellent selectivity versus PI3K-related lipid kinases, protein kinases and unrelated targets. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8248