Learn More
Selleck Chemical LLC Bindarit 5mg 130641-38-2 AF 2838
SDP
Supplier:  Selleck Chemical LLC S30325MG
Encompass_Preferred
Bindarit (AF 2838) exhibits selective inhibition against monocyte chemotactic proteins MCP-1/CCL2, MCP-3/CCL7 and MCP-2/CCL8. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3284