Learn More
TARGETMOL CHEMICALS INC BINIMETINIB 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2508200MG
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Binimetinib (ARRY-162) (MEK162 ARRY-162 ARRY-438162) is an orally available inhibitor of MEK1/2 (IC50 12 nM) in a cell-free assay. purity: 99%
Catalog No. 50-258-0769