Learn More
Selleck Chemical LLC Binimetinib
SDP
Supplier:  Selleck Chemical LLC S7007200MG
Encompass_Preferred
Binimetinib (MEK162 ARRY-162 ARRY-438162) is a potent inhibitor of MEK1/2 with IC50 of 12 nM in a cell-free assay Binimetinib induces G1 cell cycle arrest and apoptosis in human NSCLC cell lines and induces autophagy Phase 3
Catalog No. 50-000-02130