Learn More
Medchemexpress LLC Binimetinib | 606143-89-9 | C17H15BrF2N4O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY152025MG
Encompass_Preferred
Binimetinib is an oral and selective MEK1/2 inhibitor that inhibits MEK with an IC50 of 12 nM. This laboratory chemical is primarily used for the manufacture of substances.
  • Oral and selective MEK1/2 inhibitor
  • Inhibits MEK with an IC50 of 12 nM
  • Suitable for use as a laboratory chemical
  • Utilized in the manufacture of substances
Catalog No. 50-003-74127