Learn More
Selleck Chemical LLC BIO
SDP
Supplier:  Selleck Chemical LLC S71981G
Encompass_Preferred
BIO (GSK-3 Inhibitor IX 6-bromoindirubin-3-oxime 6-Bromoindirubin-3 -oxime MLS 2052) is a specific inhibitor of GSK-3 with IC50 of 5 nM for GSK-3 / in a cell-free assay shows 16-fold selectivity over CDK5 also a pan-JAK inhibitor with IC50 of 30 nM for Tyk2 BIO induces apoptosis in human melanoma cells
Catalog No. 50-000-04717