Learn More
Enzo Life Sciences Bio-11-CTP (250nmol)
SDP
Supplier:  Enzo Life Sciences ENZ42818
Encompass_Preferred
Bio-11-CTP (Biotin-11-cytidine-5’-triphosphate) can replace CTP for in vitro transcription reaction catalyzed by T3, T7 or SP6 RNA polymerases. The biotin-labeled RNA transcripts produced by these reactions are suitable for a wide range of applications such as nucleic acid hybridization, sequencing, and genome analysis. The transcription reaction produces multiple RNA copies of the DNA template(s) during a short incubation period. RNA probes offer higher target specificity and greater sensitivity than the corresponding DNA-DNA hybrids. The single-stranded RNA probes offer slectivity unavailable with double-stranded DNA probes, because they are strand-specific. Furthermore, RNA probes hybridize much more efficiently to target molecules than DNA probes because there is no self-hybridization. Alternative name: Biotin-11-cytidine-5'-triphosphate. Concentration: 10mM. Purity: ≥93% (HPLC). Formulation: Liquid. Solution in water.
Catalog No. 50-201-2543