Learn More
Selleck Chemical LLC BIO-2007817 E5771-1G
SDP
Supplier:  Selleck Chemical LLC E57711G
Encompass_Preferred
BIO-2007817 is a tetrahydropyrazolo-pyrazine and a positive allosteric modulator(PAM) that can act as a molecular glue to enhance the ability of pUb to activate Parkin with an EC50 of 150 nM It can be used in research design of activators as therapeutics for Parkinson s disease
Catalog No. 50-000-42735