Learn More
Thomas Scientific 96 Deep-Well Cover, PP
SDP
Supplier:  Thomas Scientific 9383Q05
Encompass
Expanded capability for multi-channel pipetting 2 mL capacity per well Allows for serial dilution procedures Polypropylene construction Works on most robotic instruments Optional cover Specially designed covers can be used to separate each well via a sealing ring. This reduces cross-contamination and allows for stacking the plates. Plates and covers work well with most robotic work stations including Zymark, Tecan, Hamilton, Rasys and Beckman. Side loader collars are required for side loading Beckman BioMek systems.
Catalog No. 50-145-8619