Learn More
MESA LABS BOZEMAN MFG FACILITY BIO INDCTRS 100PK
SDP
Supplier:  MESA LABS BOZEMAN MFG FACILITY 35.KCD404
Encompass
Stainless steel discs for VHP sterilization; 10e4/unit; Tyvek/Tyvek packaged; ATCC#12980
Catalog No. NC0420355