Learn More
Bio Rad Laboratories BIO-PLEX VALIDATION KIT 4.0
Supplier:  Bio Rad Laboratories 171203001
Encompass
Kit, Validation; Bio-Rad; Bio-Plex; 4.0; Includes optics validation, reporter validation, classify validation, fluidics validation bead sets for approx. 50 validation routines using Bio-Plex Manager 4.0 or 4.1 software and MCV plate III
Catalog No. 50-901-0007