Learn More
ACROBiosystems Biotinylated Rhesus Macaque CD89 (FCAR) Protein, His-AviTag, 200 μg
Supplier: ACROBiosystems CD9R82E3
Biotinylated Rhesus macaque CD89 (FcαRI) is a recombinant extracellular domain expressed in human 293 (HEK293) cells, with a C-terminal His tag and AviTag that is enzymatically biotinylated. Supplied as a 200 μg vial, it is suitable for binding and bioactivity assays such as ELISA.
- Size: 200 μg.
- Species: Rhesus macaque.
- Expression system: HEK293 cells.
- Tags and modification: C-terminal His tag and AviTag with enzymatic biotinylation.
- Sequence: amino acids Gln 22-Asn 227.
- Purity: >90% (SEC-MALS verified).
- Apparent molecular weight: migrates ~35-60 kDa on SDS-PAGE due to glycosylation.
- Applications: bioactivity ELISA and binding studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.