Learn More
Bio X Cell Bio X Cell InVivoPlus Anti-Mouse Ly6G/Ly6C (Gr-1) Monoclonal Antibody, 25 mg
Supplier: Bio X Cell BP0075R025MG
Bio X Cell InVivoPlus anti-mouse Ly6G/Ly6C (Gr-1) is a purified monoclonal antibody (clone RB6-8C5) intended for detection and in vivo depletion of Gr-1-expressing myeloid cells in mouse models. It is supplied in PBS and available in multiple sizes, including a 25 mg vial.
- Clone RB6-8C5
- Target mouse Ly6G and Ly6C (Gr-1)
- Isotype rat IgG2b, κ
- Formulation PBS, pH 7.0; no stabilizers or preservatives
- Purity ≥95%
- Endotoxin ≤0.5 EU/mg
- Purification Protein G
- Applications in vivo depletion, flow cytometry, and immunohistochemistry
- Storage store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.