Learn More
Bio X Cell Bio X Cell InVivoPlus Polyclonal Syrian Hamster IgG, 100 mg (BP0087)
Supplier: Bio X Cell BP0087R100MG
Bio X Cell InVivoPlus polyclonal Syrian hamster IgG (MPN BP0087) is a purified immunoglobulin preparation used as a non-reactive, species-matched control for experiments employing Syrian hamster antibodies. The product is formulated in PBS (pH 7.0), contains no stabilizers or preservatives, and is quality-tested for purity and endotoxin to support in vivo and in vitro applications.
- Purified from Syrian hamster serum.
- Formulated in PBS, pH 7.0.
- Contains no stabilizers or preservatives.
- Purity ≥95% by SDS-PAGE.
- Endotoxin ≤0.5 EU/mg (LAL assay).
- Available in 5 mg, 25 mg, 50 mg, and 100 mg sizes.
- Intended as a non-reactive control IgG for Syrian hamster antibodies in most in vivo and in vitro applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.