Learn More
Medchemexpress LLC BIO8898 | 98.8% | 909.13 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY122663A
Encompass_Preferred
BIO8898 is a potent inhibitor of CD40-CD154. It inhibits soluble CD40L binding to CD40-Ig with an IC50 value of 25 μM and also inhibits CD40L-induced apoptosis.
  • Potent CD40-CD154 inhibitor.
  • Inhibits soluble CD40L binding to CD40-Ig.
  • Inhibits CD40L-induced apoptosis.
Catalog No. 50-004-39060