Learn More
Agilent Technologies High Sensitivity DNA Kit
Supplier:  Agilent Technologies 50674626
Encompass
For the separation, sizing and quantification of dsDNA samples of limited abundance and sized from 50 - 7000 bp. The kit includes 10 microfluidic chips, reagents and consumables. Note: incompatible with G2938A Bioanalyzer instruments.
Catalog No. NC1738319