Learn More
SOLVENTUM Zeta Plus BC Series Filter Capsules, 2.63 inch Diameter, Pack of 4
Supplier:  SOLVENTUM 7100004078
Encompass_Preferred
This item has a minimum qty of 4 per supplier requirements.
3M designed Zeta Plus BC Series Filter Capsules with SP Series Media for biological, small molecule pharmaceutical, and food and beverage manufacturing applications where an optimized balance of throughput and particle removal efficiency is critical. These capsules, having the same media as production products, are ideal for assessing filtration media grades using low challenge fluid volumes.
Catalog No. NC0554076