Learn More
Medchemexpress LLC Biochanin A | 491-80-5 | MFCD00006839 | 98.7% | C16H12O5 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14595R10MG
Encompass_Preferred
Biochanin A (standard) is an analytical reference standard of the isoflavone Biochanin A (CAS 491-80-5) intended for use as a chromatographic and mass-spectrometry reference in qualitative and quantitative method development. Supplied as a powder, it offers high chromatographic purity and defined storage recommendations to preserve stability.
  • High purity suitable for reference and quantitative analysis.
  • Validated for use in LC, GC, and mass spectrometry workflows.
  • Available in small pack sizes for analytical testing.
  • Molecular formula C16H12O5 and molecular weight 284.26 g/mol.
  • Store as powder at low temperature to maintain stability.
Catalog No. 50-002-58438