Learn More
FOCUS BIOMOLECULES Biochemical Reagents, BML-260, JSP-1 dual specificity phosphatase inhibitor, CAS 101439-76-3, 98% by TLC/NMR (Conforms)
SDP
Supplier:  FOCUS BIOMOLECULES 105153
Encompass_Preferred
Biochemical Reagents, BML-260, JSP-1 dual specificity phosphatase inhibitor, CAS 101439-76-3, 98% by TLC/NMR (Conforms), BML-260 is a potent inhibitor of the dual specificity phosphatase JSP-1 (IC50 18 uM).1 It promotes the growth of cord blood and mobilizes peripheral blood hematopoietic stem and progenitor cells.2 It increases UCP1 expression in both brown and white adipocytes increasing mitochondrial activity and heat generation, an effect that is JSP-1 independent.3
Catalog No. NC3797295