Learn More
FOCUS BIOMOLECULES Biochemical Reagents, BRM014, BRM / BRG1 ATPase inhibitor, CAS 2270879-17-7, 98% by HPLC/NMR (Conforms)
SDP
Supplier:  FOCUS BIOMOLECULES 105244
Encompass_Preferred
Biochemical Reagents, BRM014, BRM / BRG1 ATPase inhibitor, CAS 2270879-17-7, 98% by HPLC/NMR (Conforms), BRM014 is an allosteric dual inhibitor of BRM and BRG1 (IC50less than5 nM for both). It downregulated BRM-dependent gene expression and displayed antiproliferative activity in a BRG1-mutant-lung-tumor xenograft model after oral administration. Inhibition of BRG1/BRM ATPase by BRM014 inhibits chromatin openness. For example, it reduced DNA accessibility to core regulatory factors (RUNX1, LM01 and MEIS1) at transcription factor PU.1 sites in AML cells. P583 cells formed markedly large spheroids when treated with BRM014. Cell permeable and active in vivo.
Catalog No. NC3797304