Learn More
FOCUS BIOMOLECULES Biochemical Reagents, Ionomycin Ca, Calcium Ionophore, CAS 56092-82-1, 99% by TLC/NMR (Conforms)
SDP
Supplier:  FOCUS BIOMOLECULES 102078
Encompass_Preferred
Biochemical Reagents, Ionomycin Ca, Calcium Ionophore, CAS 56092-82-1, 99% by TLC/NMR (Conforms), Polyether antibiotic and calcium ionophore isolated from the bacterium Streptomyces conglobatus.1,2
Catalog No. NC3796526