Learn More
FOCUS BIOMOLECULES Biochemical Reagents, LXG6403, Lysyl oxidase inhibitor, CAS 315705-04-5, 98% by TLC/NMR (Conforms)
SDP
Supplier:  FOCUS BIOMOLECULES 104977
Encompass_Preferred
Biochemical Reagents, LXG6403, Lysyl oxidase inhibitor, CAS 315705-04-5, 98% by TLC/NMR (Conforms), LXG6403 is a potent (IC50 1.3 uM) and selective (3.5x over LOXL2 and no inhibition of LOX1) lysyl oxidase (LOX) inhibitor. Active against LOX in multiple cell lines (IC50 less than5 uM against MDA-MB-231, HCC143, Hs-578-T, and HCC1937). LXG6403 enhanced chemosensitivity of triple negative breast cancer cells to anthracycline, doxorubicin, cisplatin, and paclitaxel. It inhibited collagen crosslinking and deposition, increased doxorubicin penetration in cells and organoids, inhibited FAK signaling, and potentiated ROS/DNA damage axis in combination with cisplatin. LXG6403 was able to overcome doxorubicin resistance in the TM01278 TNBC PDX model.
Catalog No. NC3797294