Learn More
FOCUS BIOMOLECULES Biochemical Reagents, Oligomycin complex, Mitochondrial ATPase inhibitor, CAS 1404-19-9, Mixture of Oligomycin A (67.5%) B (19.4%) and C (9.8%)/97% by TLC/NMR: (Conforms)
SDP
Supplier:  FOCUS BIOMOLECULES 10209225MG
Encompass_Preferred
Biochemical Reagents, Oligomycin complex, Mitochondrial ATPase inhibitor, CAS 1404-19-9, Mixture of Oligomycin A (67.5%) B (19.4%) and C (9.8%)/97% by TLC/NMR: (Conforms), Inhibits mitochondrial F1F0 ATP synthase1. A useful tool for decreasing cellular ATP levels2. Induces autophagy3. Stimulates lysosome acidification4. Protects against ischemic kidney in male rats5.
Catalog No. NC3802456