Learn More
FOCUS BIOMOLECULES Biochemical Reagents, PSB-KK1445, GPR18 agonist, CAS 885896-54-8, 98% by HPLC/NMR (Conforms)
SDP
Supplier:  FOCUS BIOMOLECULES 104918
Encompass_Preferred
Biochemical Reagents, PSB-KK1445, GPR18 agonist, CAS 885896-54-8, 98% by HPLC/NMR (Conforms), PSB-KK1445 is a potent (EC50 45.4 nM human/ 124 nM mouse) and highly selective (200-fold over CB1, CB2, GPR55, and GPR183) GPR18 agonist. A new chemical tool to explore the role of GPR18 in cancer and immunology.
Catalog No. NC3797290