Learn More
FOCUS BIOMOLECULES Biochemical Reagents, Troleandomycin, Macrolide antibiotic/ Cytochrome P450 inhibitor, CAS 2751-09-9, 98% by TLC/NMR (Conforms)
SDP
Supplier:  FOCUS BIOMOLECULES 10222725MG
Encompass_Preferred
Biochemical Reagents, Troleandomycin, Macrolide antibiotic/ Cytochrome P450 inhibitor, CAS 2751-09-9, 98% by TLC/NMR (Conforms), Macrolide antibiotic. Inhibits cytochrome P450 (IC50 0-5 uM), thus blocking CYP450-dependent drug oxidation.
Catalog No. NC3802457