Learn More
Medchemexpress LLC Biocytin hydrazide | 102743-85-1 | MFCD00133143 | 99.2% | C16H30N6O3S | 10MG
SDP
Supplier:  Medchemexpress LLC HY14716810MG
Encompass_Preferred
Biocytin hydrazide is a biotin-containing hydrazide reagent used to label glycoconjugates. It selectively reacts with oxidized sugar residues to introduce a biotin tag for detection by avidin-biotin methods and is supplied for research use.
  • Selectively labels sialic acids, galactose, and other aldehyde-containing sugars.
  • Introduces a biotin tag for detection with avidin-biotin methods.
  • Water-soluble and compatible with aldehyde fixation for microscopy.
  • Suitable for biochemical assays and histochemical glycoprotein detection.
  • Supplied in small research-use quantities for laboratory experiments.
  • High purity supports consistent labeling performance.
Catalog No. 50-002-58222