Learn More
Cytiva Biodyne Plus Membrane, 0.45 µm - 132 mm discs (50/pkg)
Supplier:  Cytiva 60404
Encompass
High sensitivity and low background for enhanced detection and resolution. Stands up to multiple cycles of hybridization, stripping, and reprobingHydrophilic for easy wetting. Superior performance with radioactive and non-radioactive detection systemsBiodyne A - (Amphoteric) Membrane zeta potential can be modulated by changes in pH. Ideal for single probe or multiple rehybridizations, and background reduction. Biodyne B - (Positively-charged) Pore surfaces are populated by a high density of quaternary ammonium groups. Our highest sensitivity nylon membrane for nucleic acid applications. Biodyne C - (Negatively-charged) Can be derivatized through the carboxyl groups on the pore surfaces. Biodyne Plus - (Positively-charged with an extremely high isoelectric point) With certain nonradioactive detection systems, it is more sensitive than Biodyne A while exhibiting lower background. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3250