Learn More
Dojindo Molecular Technologies Inc Biofilm Formation Assay Kit measures the amount of biofilm and drug-mediated inhibitions of biofilm formation by using crystal violet, shorten measurement time, reduce measurement variation 100 tests
SDP
Supplier:  Dojindo Molecular Technologies Inc B60110
Encompass_Preferred
Biofilms are biological aggregates consisting of microorganisms and extracellular polysaccharides that can exist in many environments. Bacterial contamination of medical devices, and infectious diseases such as caries and periodontal disease, are attributed to biofilms. The microorganisms in biofilms are highly resistant to antibiotics. Therefore, research into medicines and food ingredients that have anti-biofilm activities is a growing area. The Biofilm Formation Assay Kit measures the amount of biofilm formation and the anti-biofilm activity of samples. As the biofilm is formed on a peg layer on the lid of a microplate, multiple samples can easily be washed and measured at once without having to peel the biofilm off for each process. Dojindo offers various reagents for bacteria research. (https://www.dojindo.com/observe-bacteria-from-various-parameters/)
Catalog No. NC1933289