Learn More
Research Products International Corp BioFlsk Erlnmyr Flask Baff Bot
SDP
Supplier:  Research Products International Corp 400853
Encompass
BioFlast Cell Culture Erlenmeyer Flasks are made from a variety of disposable and autoclavable plastic. Vented screw caps (polypropylene) feature 0.2mm membranes to increase air transmission and enhance cell growth. Polycarbonated flasks and Jumbo Flasks have baffled (ribbed) bottoms to provide increased oxygenation and mixing efficiently even at lower shaking speeds. Custom mouth size conforms with the SBS standard for quick, secure connection to popular bioreactor equipment. Supplied individually wrapped and gamma sterilized (SAL 10-6 ).
RNase, DNase, Pyrogen free
Vented screw cap
Gamma sterilized
Individually wrapped
Catalog No. 50-238-8375