Learn More
Andwin Scientific BIOHAZARD BAG 12INX24IN 250CS
SDP
Supplier:  Andwin Scientific 14220082
Encompass
The Autoclavable Biohazard Bags provide extra puncture resistance and high impact strength. The bags are autoclavable to 135C (275F). This product has a clear readable label proper biohazard warnings. It is in bright red color to allow fast identif
Catalog No. NC9916166