Learn More
Sigma Aldrich Fine Chemicals Biosciences Biohazard waste container1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z3706651EA
Encompass
Biohazardous Waste Containers are ideal autoclavable polypropylene containers for secondary containment of biohazardous waste materials. Convenient one-step sterilization no need to remove bag. Port opening in cover allows one-handed disposal of materials limits exposure to infectious waste. Holds standard sized autoclave bags. Will not rust dent or puncture.
Catalog No. 50-290-6037