Learn More
MESA LABS BOZEMAN MFG FACILITY EZTest Biological Indicator, 100 pack
Supplier: MESA LABS BOZEMAN MFG FACILITY EZS/5
EZTest Biological Indicator, Contains bacterial spores on a filter-paper carrier packaged within a small, thermoplastic culture tube, EZTest steam/flash, Incubation: 24 Hours at 55deg. to 60deg.C, Population: 105 *This item is non-returnable*
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.