Learn More
Polysciences, Inc. BioMag Plus Concanavalin A, 3 mL
SDP
Supplier:  Polysciences, Inc. 860573
Encompass
Our Con A-coated BioMag Plus microparticles provide a convenient means for isolating mannosyl- and glucosyl-containing glycoproteins and polysaccharides from serum or cell lysate, or for investigating other lectin / glycan-mediated processes. The BioMag Plus magnetic particle format provides high surface area, and permits easy and efficient separations.
Catalog No. NC1358578