Learn More
Bellco Glass, Inc. Biometer Flask Only,250mL
SDP
Supplier:  Bellco Glass, Inc. 255610250
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Designed for the measurement of microbial co2 production over time. Ideal for bio-remediation studies.. MADE IN THE USA All components are autoclaveable. Flask only 250mL, 1 each.
Catalog No. 50-194-5566