Learn More
Advanced Biomatrix Printing probe for 6-well plates for BIONOVA X DLP Printer
Supplier:  Advanced Biomatrix D16110025291
Encompass_Preferred
BIONOVA X Printing Probes are compatible with the BIONOVA X 3D bioprinter from CELLINK, the only DLP based bioprinter for direct in-well printing. Available for 6, 12 and 24-well multi-well printing. Each probe comes sterile and is capable of 24-hours of continuous printing.
Catalog No. 50-225-5977