Learn More
AB Sciex LLC BIOPHASE BFS CAPILLARY 8X50CM
Supplier: AB Sciex LLC 5080123
- 8 capillary cartridge designed for sensitive electrophoretic separations
- Bare fused silica (BFS) material for enhanced performance
- Capillary count: 8
- Capillary length: 50 cm
- Compatible with BioPhase 8800 system
- Detection window located 40 cm from inlet side
- Integrated electrodes for efficient operation
- Liquid temperature control to support standardized separation temperatures
- Ensures high levels of assay repeatability
- Designed for high-throughput analysis of biotherapeutics
- Primary applications include CE-SDS, cIEF, RNA analysis, glycan characterization
- Intended for research use only
- Kit includes one cartridge with 8 x 50 cm BFS capillaries pre-installed
Description
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.