Learn More
YMC America, Inc BioProSS S10 10um 100x20mm
SDP
Supplier:  YMC America, Inc SSA0S101020WT
Encompass
YMC s BioPro IEX resins are fully scalable exhibiting consistent chromatographic behavior across all particle sizes YMC-BioPro IEX SmartSep Q/S (10 20 or 30 um) which show high resolution and recovery even at a high flow rate and high loading condition are suitable for an intermediate purification step and a polishing step of mAbs proteins peptides and oligonucleotides
Catalog No. 50-008-77725