Learn More
Electron Microscopy Sciences IP ActivFlo Biopsy II Cassettes, Lilac
SDP
Supplier:  Electron Microscopy Sciences 7007669
Encompass_Preferred
Patented "Fluidic Flow" pore design maximizes reagent exchange. IP ActivFlo Biopsy Cassettes feature innovative V-shaped 0.67 mm pores that pull fluid in and force air out. Sloped walls create a basket for small tissue management. The one-piece cassettes with 'frangible lids' have been designed for the Leica IPC Cassette Printer, with <1% printing error rate.
Color: Lilac
Catalog No. 50-192-6995