Learn More
BioSpec Products BioPulverizer, capacity 10-100mg
SDP
Supplier:  BioSpec Products 59012N
Encompass_Preferred
The BioPulverizer quickly fragments hard frozen tissue samples into tiny pieces the size of grains of sand or course powder. The primary benefit of pulverizing pieces of tissue is to optimize subsequent rapid and complete cell lysis using lytic solutions or mechanical homogenizers. For difficult tissues such as tendon, skin, and tumors it is an essential step (read more). The BioPulverizer is also used at room temperature to fragment friable, dry biomaterial such as non-oily seeds, bone and teeth.
Catalog No. NC1068180