Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp BioResolve Protein A Affinity Column, MaxPeak Premier, 3.5 μm, 3.9 x 5 mm

Supplier:  Waters Corp 186011379

Encompass_Preferred

This BioResolve Protein A Affinity Column helps analytical and process development scientists determine antibody titer measurements earlier in cell harvest. It offers up to 3x greater sensitivity for faster decision-making. MaxPeak High Performance Surface (HPS) technology reduces unwanted analyte interactions, improving peak shape and recovery. It supports simplified 2D methods for reliable titer and aggregation quantification.

  • Affinity chemistry; spherical, solid core particles
  • pH range: 2 to 10; temperature limits: 40°C
  • Maximum pressure: 6000 psi (415 Bar)
  • Particle size: 3.5 μm; dimensions: 3.9 x 5 mm
  • Compatible with HPLC, UHPLC, UPLC systems
  • Features MaxPeak Premier and High Performance Surface (HPS) technologies

Catalog No. NC3797116


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.