Learn More
Waters Corp BioResolve RP mAb Polyphenyl Column, 450Å, 2.7 µm, 3 mm X 150 mm, 1/pk
Supplier:  Waters Corp 186008950
Encompass_Preferred
BioResolve RP mAb Columns contain 2.7-μm superficially-porous-silica particles having an average pore diameter of 450Å. The particles are functionalized with a proprietary polyphenyl bonding process that was specifically designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments. This Method Validation Kit Part Number contains (3) columns from (3) different manufactured batches. Note: P/N 176004165 contains a BioResolve RP mAb Polyphenyl Column, 450A, 2.7 μm, 3mm X 100mm, 1/pkg with Intact mAb and mAb Subunit Standards
Catalog No. NC1626158