Learn More
Waters Corp BioResolve RP mAb Polyphenyl Column, 450Å, 2.7 µm, 4.6 mm X 100 mm, 1/pk with Intact mAb and mAb Subunit Standards
Supplier:  Waters Corp 176004168
Encompass_Preferred
BioResolve RP mAb Columns contain 2.7-μm superficially-porous-silica particles having an average pore diameter of 450Å. The particles are functionalized with a proprietary polyphenyl bonding process that was specifically designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments. This P/N also includes one vial of the Intact mAb Mass Check Standard (P/N 186006552) and one vial of the mAb Subunit Standard (P/N 186008927).
Catalog No. 50-152-8575