Learn More
Waters Corp BioResolve RP mAb Polyphenyl, 450Å, 2.7 μm, 3 x 50 mm
Supplier:  Waters Corp 186008948
Encompass_Preferred
This BioResolve RP mAb Polyphenyl Column is for reverse phase protein separation and analysis. It uses 2.7 μm superficially-porous-silica particles with a 450Å pore diameter, functionalized for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments.
  • Optimized for reverse phase protein separation and analysis.
  • Polyphenyl bonding for LC/LC-MS of intact IgG monoclonal antibodies and fragments.
  • Manages biotherapeutic drug analysis challenges with high throughput and sensitivity.
  • VanGuard column protection enhances lifetime and performance.
  • Addresses challenges in intact protein analysis of monoclonal antibodies and antibody drug conjugates.
  • Works with UHPLC, UPLC, and HPLC systems, supporting higher flow rates and back pressures.
  • Offers a wide and usable pH range.
  • Reduces injection-to-injection carryover.
Catalog No. 50-239-1292