Learn More
Waters Corp HPLC Column, BioResolve RP mAb Polyphenyl, 450Å, 2.7μm, 4.6 mm x 50 mm
Supplier: Waters Corp 186008953RF
The BioResolve RP mAb Polyphenyl Column is engineered for advanced reverse phase protein separation and analysis. It features 2.7 μm superficially-porous-silica particles with a 450Å pore diameter, functionalized via a proprietary polyphenyl bonding process. Optimized for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments, it includes an electronic eConnect column tag for automatic identification, tracking, and digital tracing of usage history.
- Batch qualified with a specific mAb Subunit Standard.
- Supports intact protein, mAb subunit analysis, ADC characterization, AAV, and antibody conjugates.
- Operating pH range: 2.0 to 7.0.
- Operating temperature range: 50°C to 90°C.
- Pressure stability: up to 689 bar.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.